Thermo Scientific™ 0.2 mL Strip Tubes with Caps Promotion
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip
Manufacturer:  Thermo Scientific™ AB1191PM
 View more versions of this product
Catalog No. AB1191PM
Description
Description
  • Compatible with 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips
  • Ultra-Clear cap options available for use in qPCR assays
Specifications
Specifications
Ultra Clear Cap Strips
White
Polypropylene
Certified free of human DNA, RNase, DNase, and endotoxins
For PCR and qPCR applications
10 packs of 12 strips