Please login to your online account to display your discounted pricing
Thermo Scientific™ 0.1 mL Individual UTW Tubes
Reduce incubation times during PCR with these ultra-thin wall individual PCR tubes with attached flat caps.
Manufacturer: thermo scientific™  TUC0010
 View more versions of this product
 Disclaimers
Certified free from human DNA, RNases, and DNases.
Catalog No. TUC0010
  • / Case of 960 Each
Description & Specifications
Specifications
Color Clear
Closure Color Clear
Type Tubes
Description With attached optical flat caps
UTW vessels have walls that are 0.15 mm thin where it matters the most - at the contact point between the thermal cycler block and the reactants. Thinner walls offer less thermal resistance for transferring heat from the block to the sample within the tube. This results in the ability to perform very rapid protocols by reducing incubation times during PCR.
HIghlights
  • Caps form a secure seal, yet are easy to open and close
  • Compatible with thermal cyclers accepting low-profile 0.2mL tubes
  • Ultra Thin Wall (UTW) low profile for fast PCR applications, compatible with the 24-well Piko Thermal Cycler and PikoReal Real-Time PCR System