Français
Sales & Service: 1-800-234-7437 7:30 AM - 7:30 PM EST help@fishersci.ca Français
Please login to your on-line account to display your discounted pricing
Sodium Hydroxide Solution (1N/Certified), Fisher Chemical
Sodium Hydroxide Solution
1N
Certified
0.995 to 1.005N
Volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate.
Caustic Soda Solution,
[ 1310-73-2 ] [ 7732-18-5 ]
MAP=={items=Y, details=N, specification=Y, physicalProperties=Y, safetyHandlong=Y, accesories=N, detailsSpecTab=N, isMsds=true}
  1. Specification
  2. Physical Properties
  3. Safety and Handling
Item description Catalog No. SDS Qty List Price
Poly bottle; 1L
[SODIUM HYDROX SOL 1N CERT 1L]
SS266-1
Fisher Scientific Company
No.: BPSS266-1
PolyPac; 20L
[SODIUM HYDROX SOL 1N CERT 20L]
SS266-20
Fisher Scientific Company
No.: FLSS266-20
Poly bottle; 4L
[SODIUM HYDROX SOL 1N CERT 4L]
SS266-4
Fisher Scientific Company
No.: FLSS266-4