Fisherbrand™ Isotemp Digital Dry Baths\/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.<\/p>
Applications:<\/p>
Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation<\/p>","isSciEd":"","authored":true,"skuType":"SINGLEPDP","numIssueInStd":1,"actualBonusCode":"NR","faqEnabled":false,"hazardousChargable":false,"imageGallery":[{"thumbnail":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860021 FB_Isotemp_1 BLK_front.tif-150.jpg","largeImage":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860021 FB_Isotemp_1 BLK_front.tif-650.jpg","normalImage":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860021 FB_Isotemp_1 BLK_front.tif-250.jpg"}],"chemSafety":{},"uncondensedPartNumber":"88860021","iceChargable":false,"hasRequiredAccessory":false,"image":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860021 FB_Isotemp_1 BLK_front.tif-150.jpg","requires":"Unit must be ordered with block(s)","gsaNumber":"","seokeyword":"fisher-scientific-isotemp-digital-dry-baths-block-heaters","productTitleNoGlyphs":"Fisherbrand™ Isotemp™ Digital Dry Baths\/Block Heaters","validOnRims":false,"erpProductTitle":"ISOTEMP DRY BATH 1BLOCK 120V","casNumbers":[],"seoUrl":"\/shop\/products\/fisher-scientific-isotemp-digital-dry-baths-block-heaters\/88860021","servicePlan":"","warranty":"2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.","viewAvailablePromotions":true,"shippedExpress":false,"hasAccessory":true,"leafNodeId":"2643769","returnable":false,"hasPromo":true,"standardUnitHeight":0.0,"metaKeywords":"","stdUnits":"EA","includes":"","hasServices":false,"thirdPartyNoContent":false,"alternateUnitHeight":0.0,"customerOwned":false,"alerts":"","standardUnitWeight":0.0,"numIssueInAlt":0,"standardUnitWidth":0.0,"partNumber":"88860021","materialDescription":"ISOTEMP DRY BATH 1BLOCK 120V","vendorName":"Fisherbrand™","msdsAvailable":false,"metaDescription":"Increase lab versatility","skuDifferentiator":"Single block; 100-120V","shippedDirect":false,"vendorId":"86980","usedPart":false,"specifications":{"Height (Metric)":"100 mm","Height (English)":"3.9 in.","No. of Places":"1","Length (Metric)":"288 mm","Temperature Accuracy":"≤ ± 0.5°C","Type":"Dry Bath Block","Width (English)":"7.9 in.","Warranty":"2 Year, Replacement","Plug Type":"US","Temperature Range (Metric)":"Ambient +5° to 130°C","Material":"Powder-coated Steel","Fuse":"250V 2.5A","Certifications\/Compliance":"CE, cULus, RoHS","Weight (Metric)":"2.9 kg","Timer":"0 to 99:59 min or continuous","Weight (English)":"6.39 lb.","Relative Humidity":"<80%","Includes":"Dry bath, US style plug, block lifter","Heat Up Time":"≤ 20 min. 30° to 130°C","Electrical Requirements":"100\/120 V, 50\/60 Hz","Temperature Uniformity":"≤ ± 1°C","Voltage":"100\/120 V","Length (English)":"11.3 in.","Dimensions (L x W x H)":"11.3 x 7.9 x 3.9 in. (288 x 200 x 100 mm)","Frequency":"50\/60 Hz","Controller Type":"PID","Width (Metric)":"200 mm"},"alternateUnitLength":0.0,"standardUnitLength":0.0,"disclaimers":"","certifications":"","configurable":false,"orderingAttributes":{"No. of Places":"1","Plug Type":"US"},"productTitle":"Fisherbrand™ Isotemp™ Digital Dry Baths\/Block Heaters","accessoriesUrl":"\/store1\/products\/accessories\/88860021","discontinued":false,"hazardousPackingCharge":false,"storage":""}} , {"productRules":{"hasCrossRef":false,"message":"","suspicious":false,"viewable":true,"buyable":true,"showPrice":true,"noStockOrderDisallow":false,"mustUseMFShipToAddress":false,"partFound":true,"preferred":false,"userType":"G","returnable":true},"priceAndAvailability":[{"availabilityFetched":false,"displayListPrice":false,"showQuantity":false,"hasBreakPrice":false,"whiteListFlag":false,"shippedDirect":false,"displayUnitString":"chaque","uomType":"STD","hasDiscount":false,"validOnRims":false,"hasDynamicPrice":false,"price":"$2,342.73","stockedItem":false,"listPrice":"$2,342.73","uom":"CH","honorWebPrice":false,"quantity":"1","discontinued":false,"unformattedPrice":0.0,"pricingDisplayEU":false,"shippedExpress":false}],"productDetails":{"cdcType":"1","suspicious":false,"vendorRoutingFlag":0,"familyProductUrl":"\/shop\/products\/fisher-scientific-isotemp-digital-dry-baths-block-heaters\/p-4933697","alternateUnitWidth":0.0,"franchiseNoContent":false,"productDescription":"ISOTEMP DRY BATH 4BLOCK 120V","alternateUnitWeight":0.0,"fisherItem":false,"compliance":"","productCompareEnabled":false,"numStdInAlt":0,"userType":"G","productType":"21","orderInfo":"","vendorPartNumber":"88860023","productFeatures":"
Fisherbrand™ Isotemp Digital Dry Baths\/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.<\/p>
Applications:<\/p>
Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation<\/p>","isSciEd":"","authored":true,"skuType":"SINGLEPDP","numIssueInStd":1,"actualBonusCode":"NR","faqEnabled":false,"hazardousChargable":false,"imageGallery":[{"thumbnail":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860023_4 BLK_front.tif-150.jpg","largeImage":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860023_4 BLK_front.tif-650.jpg","normalImage":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860023_4 BLK_front.tif-250.jpg"}],"chemSafety":{},"uncondensedPartNumber":"88860023","iceChargable":false,"hasRequiredAccessory":false,"image":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860023_4 BLK_front.tif-150.jpg","requires":"Unit must be ordered with block(s)","gsaNumber":"","seokeyword":"fisher-scientific-isotemp-digital-dry-baths-block-heaters","productTitleNoGlyphs":"Fisherbrand™ Isotemp™ Digital Dry Baths\/Block Heaters","validOnRims":false,"erpProductTitle":"ISOTEMP DRY BATH 4BLOCK 120V","casNumbers":[],"seoUrl":"\/shop\/products\/fisher-scientific-isotemp-digital-dry-baths-block-heaters\/88860023","servicePlan":"","warranty":"2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.","viewAvailablePromotions":true,"shippedExpress":false,"hasAccessory":true,"leafNodeId":"2643769","returnable":false,"hasPromo":true,"standardUnitHeight":0.0,"metaKeywords":"","stdUnits":"EA","includes":"","hasServices":false,"thirdPartyNoContent":false,"alternateUnitHeight":0.0,"customerOwned":false,"alerts":"","standardUnitWeight":0.0,"numIssueInAlt":0,"standardUnitWidth":0.0,"partNumber":"88860023","materialDescription":"ISOTEMP DRY BATH 4BLOCK 120V","vendorName":"Fisherbrand™","msdsAvailable":false,"metaDescription":"Increase lab versatility","skuDifferentiator":"Four blocks; 100-120V","shippedDirect":false,"vendorId":"86980","usedPart":false,"specifications":{"Height (Metric)":"100 mm","Height (English)":"3.9 in.","No. of Places":"4","Length (Metric)":"450 mm","Temperature Accuracy":"≤ ± 0.5°C","Type":"Dry Bath Block","Width (English)":"7.9 in.","Warranty":"2 Year, Replacement","Plug Type":"US","Temperature Range (Metric)":"Ambient +5° to 130°C","Material":"Powder-coated Steel","Fuse":"250V 2.5A","Certifications\/Compliance":"CE, cULus, RoHS","Weight (Metric)":"4.7 kg","Timer":"0 to 99:59 min or continuous","Weight (English)":"10.36 lb.","Relative Humidity":"<80%","Includes":"Dry bath, US style plug, block lifter","Heat Up Time":"≤ 20 min. 30° to 130°C","Electrical Requirements":"100\/120 V, 50\/60 Hz","Temperature Uniformity":"≤ ± 1°C","Voltage":"100\/120 V","Length (English)":"17.7 in.","Dimensions (L x W x H)":"17.7 x 7.9 x 3.9 in. (450 x 200 x 100 mm)","Frequency":"50\/60 Hz","Controller Type":"PID","Width (Metric)":"200 mm"},"alternateUnitLength":0.0,"standardUnitLength":0.0,"disclaimers":"","certifications":"","configurable":false,"orderingAttributes":{"No. of Places":"4","Plug Type":"US"},"productTitle":"Fisherbrand™ Isotemp™ Digital Dry Baths\/Block Heaters","accessoriesUrl":"\/store1\/products\/accessories\/88860023","discontinued":false,"hazardousPackingCharge":false,"storage":""}} , {"productRules":{"hasCrossRef":false,"message":"","suspicious":false,"viewable":true,"buyable":true,"showPrice":true,"noStockOrderDisallow":false,"mustUseMFShipToAddress":false,"partFound":true,"preferred":false,"userType":"G","returnable":true},"priceAndAvailability":[{"availabilityFetched":false,"displayListPrice":false,"showQuantity":false,"hasBreakPrice":false,"whiteListFlag":false,"shippedDirect":false,"displayUnitString":"chaque","uomType":"STD","hasDiscount":false,"validOnRims":false,"hasDynamicPrice":false,"price":"$1,937.48","stockedItem":false,"listPrice":"$1,937.48","uom":"CH","honorWebPrice":false,"quantity":"1","discontinued":false,"unformattedPrice":0.0,"pricingDisplayEU":false,"shippedExpress":false}],"productDetails":{"cdcType":"1","suspicious":false,"vendorRoutingFlag":0,"familyProductUrl":"\/shop\/products\/fisher-scientific-isotemp-digital-dry-baths-block-heaters\/p-4933697","alternateUnitWidth":0.0,"franchiseNoContent":false,"productDescription":"ISOTEMP DRY BATH 2BLOCK 120V","alternateUnitWeight":0.0,"fisherItem":false,"compliance":"","productCompareEnabled":false,"numStdInAlt":0,"userType":"G","productType":"21","orderInfo":"","vendorPartNumber":"88860022","productFeatures":"
Fisherbrand™ Isotemp Digital Dry Baths\/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.<\/p>
Applications:<\/p>
Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation<\/p>","isSciEd":"","authored":true,"skuType":"SINGLEPDP","numIssueInStd":1,"actualBonusCode":"NR","faqEnabled":false,"hazardousChargable":false,"imageGallery":[{"thumbnail":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860022_2 BLK_front.tif-150.jpg","largeImage":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860022_2 BLK_front.tif-650.jpg","normalImage":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860022_2 BLK_front.tif-250.jpg"}],"chemSafety":{},"uncondensedPartNumber":"88860022","iceChargable":false,"hasRequiredAccessory":false,"image":"https:\/\/assets.fishersci.com\/TFS-Assets\/CCG\/product-images\/88860022_2 BLK_front.tif-150.jpg","requires":"Unit must be ordered with block(s)","gsaNumber":"","seokeyword":"fisher-scientific-isotemp-digital-dry-baths-block-heaters","productTitleNoGlyphs":"Fisherbrand™ Isotemp™ Digital Dry Baths\/Block Heaters","validOnRims":false,"erpProductTitle":"ISOTEMP DRY BATH 2BLOCK 120V","casNumbers":[],"seoUrl":"\/shop\/products\/fisher-scientific-isotemp-digital-dry-baths-block-heaters\/88860022","servicePlan":"","warranty":"2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.","viewAvailablePromotions":true,"shippedExpress":false,"hasAccessory":true,"leafNodeId":"2643769","returnable":false,"hasPromo":true,"standardUnitHeight":0.0,"metaKeywords":"","stdUnits":"EA","includes":"","hasServices":false,"thirdPartyNoContent":false,"alternateUnitHeight":0.0,"customerOwned":false,"alerts":"","standardUnitWeight":0.0,"numIssueInAlt":0,"standardUnitWidth":0.0,"partNumber":"88860022","materialDescription":"ISOTEMP DRY BATH 2BLOCK 120V","vendorName":"Fisherbrand™","msdsAvailable":false,"metaDescription":"Increase lab versatility","skuDifferentiator":"Dual block; 100-120V","shippedDirect":false,"vendorId":"86980","usedPart":false,"specifications":{"Height (Metric)":"100 mm","Height (English)":"3.9 in.","No. of Places":"2","Length (Metric)":"318 mm","Temperature Accuracy":"≤ ± 0.5°C","Type":"Dry Bath Block","Width (English)":"7.9 in.","Warranty":"2 Year, Replacement","Plug Type":"US","Temperature Range (Metric)":"Ambient +5° to 130°C","Material":"Powder-coated Steel","Fuse":"250V 5A","Certifications\/Compliance":"CE, cULus, RoHS","Weight (Metric)":"3.3 kg","Timer":"0 to 99:59 min or continuous","Weight (English)":"7.28 lb.","Relative Humidity":"<80%","Includes":"Dry bath, US style plug, block lifter","Heat Up Time":"≤ 20 min. 30° to 130°C","Electrical Requirements":"100\/120 V, 50\/60 Hz","Temperature Uniformity":"≤ ± 1°C","Voltage":"100\/120 V","Length (English)":"12.5 in.","Dimensions (L x W x H)":"12.5 x 7.9 x 3.9 in. (318 x 200 x 100 mm)","Frequency":"50\/60 Hz","Controller Type":"PID","Width (Metric)":"200 mm"},"alternateUnitLength":0.0,"standardUnitLength":0.0,"disclaimers":"","certifications":"","configurable":false,"orderingAttributes":{"No. of Places":"2","Plug Type":"US"},"productTitle":"Fisherbrand™ Isotemp™ Digital Dry Baths\/Block Heaters","accessoriesUrl":"\/store1\/products\/accessories\/88860022","discontinued":false,"hazardousPackingCharge":false,"storage":""}} ], "assetInfo":{ }, "userType": "G", "message":"Success" }, "author":{ "items":[ { "CQProductImage" : "", "CQProductSpec" : "", "CQProductInfo" : "", "CQProductTitle" : "", "CQProductImageAlt" : "" } , { "CQProductImage" : "", "CQProductSpec" : "", "CQProductInfo" : "", "CQProductTitle" : "", "CQProductImageAlt" : "" } , { "CQProductImage" : "", "CQProductSpec" : "", "CQProductInfo" : "", "CQProductTitle" : "", "CQProductImageAlt" : "" } ] } }