Moisture Balances
Mettler Toledo™ HC103 Halogen Moisture Analyzer
For routine moisture determination in laboratory and production. Mettler Toledo HC103 Halogen Moisture Analyzer measures moisture content in minutes, enabling fast response times for quality control and in-process control. A large color touchscreen, graphical user guidance and real-time drying curve make the HC103 easy to use, even for untrained operators.
Sartorius MA35 Moisture Analyzers
Electronic moisture analyzer with user friendly, rugged design
Sartorius MA160 Moisture Analyzer
Perform fast and precise analysis on a diverse range of product samples with maximum flexibility. Sartorius MA160 Moisture Analyzer uses the thermogravimetric method to determine the moisture content of liquid, pasty and solid substances conveniently, reliably and in minimum time.
Mettler Toledo™ HE73 Halogen Moisture Analyzer
The Mettler Toledo HE73 Halogen Moisture Analyzer is a robust, reliable and cost-effective solution for determining moisture content of your products.
Ohaus™ MB27 Moisture Analyzer
Get dependable, accurate results for a wide variety of moisture analysis applications. The Ohaus MB27 Moisture Analyzer combines high quality and durable construction into a sleek, compact design.
Sartorius MA37 Moisture Analyzer
For quick and reliable determination of the moisture content of liquid, pasty and solid substances using the thermogravimetric method.
Mettler Toledo™ HE53 Halogen Moisture Analyzer
The Mettler Toledo HE53 Halogen Moisture Analyzer is a robust, reliable and cost-effective solution for determining moisture content of your products.
Mettler Toledo™ Halogen HX204 Moisture Analyzer
The Mettler Toledo™ HX204 Halogen Moisture Analyzer offers best-in-class measurement performance for any sample type with high-resolution weigh cell and innovative hanging weighing pan design. Designed for compliance with smart features such as user management and 'no results on instrument' option. Network connectivity and versatile reporting options make it an ideal instrument for any data management needs.
OHAUS™ MB23 / MB25 Basic Moisture Analyzers
High-value performance for all budgets. OHAUS MB23 / MB25 Basic Moisture Analyzer comes with choice of infrared or halogen heating and with precisions of 0.1% and 0.05%.
Ohaus™ MB90 Moisture Analyzer
Get both speed and precision in one moisture analyzer. The Ohaus MB90 uses halogen heating which distributes heat uniformly over the sample and speeds up the drying process, delivering fast and precise measurements. Designed for tool-free cleaning, the components can be easily removed for quick and easy maintenance. Operation is intuitive with the icon-driven menu navigation on the touchscreen display.
Ohaus™ MB120 Moisture Analyzer
Test more samples in less time. The Ohaus MB120 Moisture Analyzer delivers functionality and precision that you can rely on, and is easy to operate and clean. Advanced features such as a temperature guide help to analyze optimum drying temperature, and methods and results storage allows statistical analysis of measurements.
Mettler Toledo™ HS153 Halogen Moisture Analyzer
The Mettler Toledo™ HS153 Moisture Analyzer offers accurate results for numerous sample types with high-performance weigh cell and innovative hanging weighing pan design. Compliance features such as user management, combined with network connectivity and versatile reporting options make it an ideal instrument for any data management needs.
CSC Scientific Scale Lamp for Moisture Balance 26680-000
For use with torsion-type 115V moisture balances.
  spinner