accessibility menu, dialog, popup

UserName

CAS RN 996-23-6

OHOHOOHOHOCa

CAS RN 996-23-6

IUPAC Name: bis(2-hydroxyacetic acid) calcium
Synonyms: bis(glycolic acid) calcium
Molecular Weight (g/mol): 192.18
Molecular Formula: C4H8CaO6
InChi Key: UPYDGZJYILCHSK-UHFFFAOYSA-N
SMILES: [Ca].OCC(O)=O.OCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results