man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 887584-84-1

FFOHOCl

CAS RN 887584-84-1

IUPAC Name: 6-chloro-2,3-difluorobenzoic acid
Synonyms: 6-chloro-2,3-difluorobenzoic acid
Molecular Weight (g/mol): 192.55
Molecular Formula: C7H3ClF2O2
InChi Key: MJWXJSUYTVBLJU-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)