man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 850568-65-9

BNOOH3CH3CCH3OCH3HOHO

CAS RN 850568-65-9

IUPAC Name: {1-[(tert-butoxy)carbonyl]-6-methoxy-1H-indol-2-yl}boronic acid
Synonyms: 1-(tert-butoxycarbonyl)-6-methoxyindol-2-ylboronic acid
Molecular Weight (g/mol): 291.11
Molecular Formula: C14H18BNO5
InChi Key: ATIJYIUOXDCYTN-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(B(O)O)N(C(=O)OC(C)(C)C)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results