accessibility menu, dialog, popup

UserName

CAS RN 83164-33-4

FNHONOFFFF

CAS RN 83164-33-4

IUPAC Name: N-(2,4-difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxamide
Synonyms: diflufenican
Molecular Weight (g/mol): 394.3
Molecular Formula: C19H11F5N2O2
InChi Key: WYEHFWKAOXOVJD-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC(=O)C2=CC=CN=C2OC2=CC=CC(=C2)C(F)(F)F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results