man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 772-49-6

ClOFFF

CAS RN 772-49-6

IUPAC Name: 1-chloro-3-(trifluoromethoxy)benzene
Synonyms: 1-chloro-3-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 196.55
Molecular Formula: C7H4ClF3O
InChi Key: OLDJEKBXICPMAS-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results