accessibility menu, dialog, popup

UserName

CAS RN 722544-51-6

CH3NONNNHNHNONHFOH

CAS RN 722544-51-6

IUPAC Name: 2-{3-[(7-{3-[ethyl(2-hydroxyethyl)amino]propoxy}quinazolin-4-yl)amino]-1H-pyrazol-5-yl}-N-(3-fluorophenyl)acetamide
Synonyms: 2-{5-[(7-{3-[ethyl(2-hydroxyethyl)amino]propoxy}quinazolin-4-yl)amino]-2H-pyrazol-3-yl}-N-(3-fluorophenyl)acetamide
Molecular Weight (g/mol): 507.57
Molecular Formula: C26H30FN7O3
InChi Key: QYZOGCMHVIGURT-UHFFFAOYSA-N
SMILES: CCN(CCO)CCCOC1=CC=C2C(NC3=NNC(CC(=O)NC4=CC(F)=CC=C4)=C3)=NC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results