accessibility menu, dialog, popup

UserName

CAS RN 6628-79-1

CH3HOOCH3O

CAS RN 6628-79-1

IUPAC Name: 3-methyl-4-oxopentanoic acid
Synonyms: 3-methyl-4-oxopentanoic acid
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: NFIWUVRBASXMGK-UHFFFAOYNA-N
SMILES: CC(CC(O)=O)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results