accessibility menu, dialog, popup

UserName

CH3OH3CO

CAS RN 5726-19-2

IUPAC Name: 2-methylcyclohexyl acetate
Synonyms: 2-methylcyclohexyl acetate
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: AKIIJALHGMKJEJ-UHFFFAOYNA-N
SMILES: CC1CCCCC1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results