accessibility menu, dialog, popup

UserName

CAS RN 39450-01-6

H3CONOONOOOCH3OOPOHOHOHO

CAS RN 39450-01-6

IUPAC Name: {[6-(1-hydroxyethyl)-4-methyl-2-{[(4-nitrophenyl)methoxy]carbonyl}-7-oxo-3,4-diphenoxy-1-azabicyclo[3.2.0]hept-2-en-5-yl]oxy}phosphonic acid
Synonyms: [6-(1-hydroxyethyl)-4-methyl-2-{[(4-nitrophenyl)methoxy]carbonyl}-7-oxo-3,4-diphenoxy-1-azabicyclo[3.2.0]hept-2-en-5-yl]oxyphosphonic acid
Molecular Weight (g/mol): 626.51
Molecular Formula: C29H27N2O12P
InChi Key: OUPAKHMWUJJXOM-UHFFFAOYNA-N
SMILES: CC(O)C1C(=O)N2C(C(=O)OCC3=CC=C(C=C3)[N+]([O-])=O)=C(OC3=CC=CC=C3)C(C)(OC3=CC=CC=C3)C12OP(O)(O)=O
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results