accessibility menu, dialog, popup

UserName

CAS RN 3558-60-9

H3CO

CAS RN 3558-60-9

IUPAC Name: (2-methoxyethyl)benzene
Synonyms: methyl phenethyl ether
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: CQLYXIUHVFRXLT-UHFFFAOYSA-N
SMILES: COCCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results