CAS RN 145149-48-0
IUPAC Name:
tert-butyl N-(1-hydroxy-3-phenylpropan-2-yl)carbamate
Synonyms:
tert-butyl N-(1-hydroxy-3-phenylpropan-2-yl)carbamate
Molecular Weight (g/mol):
251.33
Molecular Formula:
C14H21NO3
InChi Key:
LDKDMDVMMCXTMO-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CO)CC1=CC=CC=C1
1
–
1
of
1
results
1
N-(tert-Butoxycarbonyl)-DL-phenylalaninol 96.0+%, TCI America™
CAS: 145149-48-0 Molecular Formula: C14H21NO3 Molecular Weight (g/mol): 251.326 MDL Number: MFCD01457313 InChI Key: LDKDMDVMMCXTMO-UHFFFAOYSA-N Synonym: N-Boc-DL-phenylalaninol, 2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol PubChem CID: 545866 IUPAC Name: tert-butyl N-(1-hydroxy-3-phenylpropan-2-yl)carbamate SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CO