man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 139756-02-8

CH3NNH2NOH2NCH3

CAS RN 139756-02-8

IUPAC Name: 4-amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
Synonyms: 4-amino-2-methyl-5-propylpyrazole-3-carboxamide
Molecular Weight (g/mol): 182.23
Molecular Formula: C8H14N4O
InChi Key: PZMXDLWWQHYXGY-UHFFFAOYSA-N
SMILES: CCCC1=NN(C)C(C(N)=O)=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results