man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 137218-25-8

FFFON

CAS RN 137218-25-8

IUPAC Name: 2-[2-(trifluoromethoxy)phenyl]acetonitrile
Synonyms: 2-[2-(trifluoromethoxy)phenyl]acetonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: OIKWCWVMUBCXJM-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results