man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 131401-55-3

H3CNCH3OFF

CAS RN 131401-55-3

IUPAC Name: N,N-diethyl-2,5-difluorobenzamide
Synonyms: N,N-diethyl-2,5-difluorobenzamide
Molecular Weight (g/mol): 213.23
Molecular Formula: C11H13F2NO
InChi Key: JEEJMEMWBHLCKZ-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=C(F)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results