accessibility menu, dialog, popup

Test

CAS RN 127425-73-4

CAS RN 127425-73-4

IUPAC Name: 1-bromo-4-(bromomethyl)-2-fluorobenzene
Synonyms: 1-bromo-4-(bromomethyl)-2-fluorobenzene
Molecular Weight (g/mol): 267.92
Molecular Formula: C7H5Br2F
InChi Key: GMVNLHLMJSMARX-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results