man

Test

2017-Fisher-Scientific-Logo
Products
Main Menu
Documents and Certificates

CAS RN 1261779-40-1

ClNOFFF

CAS RN 1261779-40-1

IUPAC Name: 2-chloro-6-(trifluoromethoxy)benzonitrile
Synonyms: 2-chloro-6-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 221.56
Molecular Formula: C8H3ClF3NO
InChi Key: NTRGETPKMRGTRQ-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(C#N)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)