CAS RN 123409-00-7
CAS RN 123409-00-7
IUPAC Name:
N-[1-(4-{[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3-phenoxyphenyl)ethyl]acetamide formic acid tritetracontakis(methane) octaamine pentadecahydrate dihydrochloride
Synonyms:
N-[1-(4-{[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3-phenoxyphenyl)ethyl]acetamide formic acid tritetracontakis(methane) octaamine pentadecahydrate dihydrochloride
Molecular Weight (g/mol):
1632.72
Molecular Formula:
C66H257Cl2N9O24
InChi Key:
ARNOCQCQWLXLRG-UHFFFAOYNA-N
SMILES:
C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.N.N.N.N.N.N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.Cl.Cl.OC=O.CC(NC(C)=O)C1=CC(OC2=CC=CC=C2)=C(OC2CC(O)C(O)C(CO)O2)C=C1
1
–
1
of
1
results
1
Vancomycin Hydrochloride MP Biomedicals
CAS: 123409-00-7 Molecular Formula: C66H257Cl2N9O24 Molecular Weight (g/mol): 1632.721 InChI Key: ARNOCQCQWLXLRG-UHFFFAOYSA-N PubChem CID: 44134997 IUPAC Name: azane;N-[1-[4-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-phenoxyphenyl]ethyl]acetamide;formic acid;methane;pentadecahydrate;dihydrochloride SMILES: C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.CC(C1=CC(=C(C=C1)OC2CC(C(C(O2)CO)O)O)OC3=CC=CC=C3)NC(=O)C.C(=O)O.N.N.N.N.N.N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.Cl.Cl