accessibility menu, dialog, popup

Test

CAS RN 120278-07-1

NNH2OO

CAS RN 120278-07-1

IUPAC Name: benzyl 4-aminopiperidine-1-carboxylate
Synonyms: benzyl 4-aminopiperidine-1-carboxylate
Molecular Weight (g/mol): 234.3
Molecular Formula: C13H18N2O2
InChi Key: YYIQGSYCCNQAGV-UHFFFAOYSA-N
SMILES: NC1CCN(CC1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results