CAS RN 107264-09-5
CAS RN 107264-09-5
IUPAC Name:
1-fluoropyridin-1-ium
Synonyms:
1-fluoropyridin-1-ium
Molecular Weight (g/mol):
98.1
Molecular Formula:
C5H5FN
InChi Key:
UWDCUCCPBLHLTI-UHFFFAOYSA-N
SMILES:
F[N+]1=CC=CC=C1
1
–
1
of
1
results
1
1-Fluoropyridinium tetrafluoroborate, 95+%, Thermo Scientific Chemicals
CAS: 107264-09-5 Molecular Formula: C5H5FN Molecular Weight (g/mol): 98.10 MDL Number: MFCD00153176 InChI Key: UWDCUCCPBLHLTI-UHFFFAOYSA-N Synonym: 1-fluoropyridinium tetrafluoroborate,n-fluoropyridinium tetrafluoroborate,1-fluoropyridin-1-ium tetrafluoroborate,acmc-209tq2,ksc496g3f PubChem CID: 2733303 IUPAC Name: 1-fluoropyridin-1-ium;tetrafluoroborate SMILES: F[N+]1=CC=CC=C1