CAS RN 10250-48-3
CAS RN 10250-48-3
IUPAC Name:
methyl 3,3-dimethylbutanoate
Synonyms:
methyl 3,3-dimethylbutanoate
Molecular Weight (g/mol):
130.19
Molecular Formula:
C7H14O2
InChi Key:
DXBOTVWRXLQVMG-UHFFFAOYSA-N
SMILES:
COC(=O)CC(C)(C)C
1
–
1
of
1
results
1
Methyl tert-butylacetate, 98%, Thermo Scientific Chemicals
CAS: 10250-48-3 Molecular Formula: C7H14O2 Molecular Weight (g/mol): 130.187 MDL Number: MFCD00043646 InChI Key: DXBOTVWRXLQVMG-UHFFFAOYSA-N Synonym: methyl tert-butylacetate,methyl 3,3-dimethylbutyrate,butanoic acid, 3,3-dimethyl-, methyl ester,butyric acid, 3,3-dimethyl-, methyl ester,methyl t-butylacetate,acmc-1c3ge,methyl-tert-butyl acetate,3,3-dimethylbutyric acid methyl ester,3,3-dimethyl-butyric acid methyl ester PubChem CID: 66298 IUPAC Name: methyl 3,3-dimethylbutanoate SMILES: CC(C)(C)CC(=O)OC