man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1000339-54-7

H3COFFFO

CAS RN 1000339-54-7

IUPAC Name: 2-methoxy-3-(trifluoromethyl)benzaldehyde
Synonyms: 2-methoxy-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: XBRULUWQNDWLMN-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results