Learn More
Zirconium, plasma standard solution, Specpure™ Zr 1000μg/mL, Thermo Scientific Chemicals
$119.32 - $119.32
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA13875AE
|
Thermo Scientific Chemicals
013875AE |
100 mL |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
MFCD00011303 |
Specifications
ZrCl2O in 5% HCl | |
MFCD00011303 | |
Specpure™ | |
Zirconium plasma standard solution, Zr 1000μg/mL |
100 mL | |
UN1789 | |
Liquid |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only