Learn More
Zincon monosodium salt, Thermo Scientific Chemicals
$58.96 - $664.73
Chemical Identifiers
CAS | 62625-22-3 |
---|---|
Molecular Formula | C20H16N4NaO6S |
Molecular Weight (g/mol) | 463.42 |
MDL Number | MFCD00064385 |
InChI Key | IABRINWJUBAIIE-JJECXDOKSA-N |
PubChem CID | 131856391 |
IUPAC Name | 2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium |
SMILES | C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2425603
|
Thermo Scientific Chemicals
B2425603 |
1 g |
N/A
|
|
|||||
AAB2425606
|
Thermo Scientific Chemicals
B2425606 |
5 g |
N/A
|
|
|||||
AAB2425614
|
Thermo Scientific Chemicals
B2425614 |
25 g |
N/A
|
|
|||||
Description
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsZincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.
Solubility
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
62625-22-3 | |
463.42 | |
IABRINWJUBAIIE-JJECXDOKSA-N | |
2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium |
C20H16N4NaO6S | |
MFCD00064385 | |
131856391 | |
C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na] |
Specifications
62625-22-3 | |
C20H16N4NaO6S | |
1 g | |
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents. | |
C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na] | |
463.42 | |
462.42 |
∽203°C (decomposition) | |
MFCD00064385 | |
3872699 | |
IABRINWJUBAIIE-JJECXDOKSA-N | |
2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium | |
131856391 | |
Zincon monosodium salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 263-651-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only