missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc Sulfate, 0.0500 M (M/20), Ricca Chemical
For EDTA back titrations
$157.16 - $263.28
Chemical Identifiers
CAS | 7446-20-0 |
---|---|
Molecular Formula | H14O11SZn |
Molecular Weight (g/mol) | 287.54 |
MDL Number | MFCD00149894 |
InChI Key | RZLVQBNCHSJZPX-UHFFFAOYSA-L |
Synonym | zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o |
PubChem CID | 62640 |
ChEBI | CHEBI:32312 |
IUPAC Name | zinc(2+) heptahydrate sulfate |
SMILES | O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
9660-16
|
Ricca Chemical Company
966016 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
9660-32
|
Ricca Chemical Company
966032 |
1 L | Natural Poly Bottle |
N/A
|
|
||||
13-622-622
|
Ricca Chemical Company
96601 |
4 L | Natural Poly Bottle |
N/A
|
|
||||
9660-5
|
Ricca Chemical Company
96605 |
20 L | Cubitainer |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7446-20-0 | |
287.54 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
62640 | |
zinc(2+) heptahydrate sulfate |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
CHEBI:32312 | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Specifications
7446-20-0 | |
MFCD00149894 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
zinc(2+) heptahydrate sulfate | |
62640 | |
0.0500M (M/20) | |
500 mL |
H14O11SZn | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
287.54 | |
CHEBI:32312 | |
Natural Poly Bottle | |
Zinc Sulfate |