missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc chromate, Thermo Scientific Chemicals

$149.71 - $538.51

Chemical Identifiers

CAS 13530-65-9
Molecular Formula CrO4Zn
Molecular Weight (g/mol) 181.37
MDL Number MFCD00036281
InChI Key NDKWCCLKSWNDBG-UHFFFAOYSA-N
Synonym zinc chromate, zinc chrome, zinc chromate c, zinc chromate o, zinc chromate t, zinc chromate z, zinc chromate am, zincro zto, basic zinc chromate, zinc chrome yellow
PubChem CID 26089
IUPAC Name zinc;dioxido(dioxo)chromium
SMILES [Zn++].[O-][Cr]([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1817830
View Documents Promotion Details
Thermo Scientific Chemicals
A1817830
250 g
Each for $149.71
 
AAA181780B
View Documents Promotion Details
Thermo Scientific Chemicals
A181780B
1000 g
Each for $538.51
 
Description

Description

Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

13530-65-9
181.37
NDKWCCLKSWNDBG-UHFFFAOYSA-N
26089
[Zn++].[O-][Cr]([O-])(=O)=O
CrO4Zn
MFCD00036281
zinc chromate, zinc chrome, zinc chromate c, zinc chromate o, zinc chromate t, zinc chromate z, zinc chromate am, zincro zto, basic zinc chromate, zinc chrome yellow
zinc;dioxido(dioxo)chromium

Specifications

13530-65-9
CrO4Zn
UN3077
zinc chromate, zinc chrome, zinc chromate c, zinc chromate o, zinc chromate t, zinc chromate z, zinc chromate am, zincro zto, basic zinc chromate, zinc chrome yellow
NDKWCCLKSWNDBG-UHFFFAOYSA-N
zinc;dioxido(dioxo)chromium
26089
Zinc chromate
3.4
MFCD00036281
250 g
Insoluble in water.
[Zn++].[O-][Cr]([O-])(=O)=O
181.37
181.37
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c

H302-H317-H350i

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 236-878-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only