missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Z-Leu-Val-Gly Diazomethyl Ketone
Supplier: Thermo Scientific Chemicals J64731MB
Description
Mimics part of the human cysteine proteinase-binding center; an irreversible inhibitor of human cysteine proteinase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Mimics part of the human cysteine proteinase-binding center
- An irreversible inhibitor of human cysteine proteinase
Specifications
25 mg | |
-30°C to -10°C | |
119670-30-3 | |
445.52g/mol | |
Signal Transduction Reagent | |
ZLDBPOCJCXPVND-OALUTQOASA-N | |
benzyl N-[(1S)-1-{[(2S)-2-[(3-diazo-2-oxopropyl)amino]-3-methylbutanoyl]carbamoyl}-3-methylbutyl]carbamate |
White to Off-white | |
Mimics part of the human cysteine proteinase-binding center; an irreversible inhibitor of human cysteine proteinase | |
C22H31N5O5 | |
C-3455 | |
Powder | |
CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)NC(=O)[C@@H](NCC(=O)C=[N+]=[N-])C(C)C | |
445.52 |
Chemical Identifiers
119670-30-3 | |
445.52 | |
C-3455 | |
CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)NC(=O)[C@@H](NCC(=O)C=[N+]=[N-])C(C)C |
C22H31N5O5 | |
ZLDBPOCJCXPVND-OALUTQOASA-N | |
benzyl N-[(1S)-1-{[(2S)-2-[(3-diazo-2-oxopropyl)amino]-3-methylbutanoyl]carbamoyl}-3-methylbutyl]carbamate |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only