missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Whitney™ Medical Solutions Bench-Top Keeper™ Biohazard Container
Economical point-of-use alternative to hard-sided sharps containers for disposal of contaminated Petri dishes, plastic assay vials, analyzer cups and other non-sharp waste
$584.25 - $615.44
Specifications
Material | Paperboard |
---|---|
Product Type | Benchtop Keeper |
Disposable | Yes |
Autoclavable | Autoclavable |
Includes | Self-locking tops |
Catalog Number | Mfr. No. | Height (Metric) | Length (Metric) | Size | Width (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (Metric) | Length (Metric) | Size | Width (Metric) | Price | Quantity | ||
14-375-265
|
Whitney™ Medical Solutions
BH2002 |
18 cm | 10 cm | Small | 10 cm |
Case of 100 Each for $584.25
|
|
||
14-375-266
|
Whitney™ Medical Solutions
BH2003 |
25 cm | 15 cm | Large | 15 cm |
Case of 50 Each for $615.44
|
|
||
Description
- Made of sturdy paperboard with an internal plastic lining and webbed corners to contain moderate amounts of fluids
- Self-locking tops keep contents safely enclosed
- Small Keepers fold out to 4L × 4W × 7 in.H (10 × 10 × 18cm) and accommodate up to 1600cc of material
- Large Keepers fold out to 6L × 6W × 10 in.H (15 × 15 × 25cm) and accommodate up to 5000cc of material
- Autoclavable
- Made in USA
Specifications
Paperboard | |
Yes | |
Self-locking tops | |
Storing contaminated lab waste, disposal of contaminated petri dishes, plastic assay vials, analyzer cups and other non-sharp waste |
Benchtop Keeper | |
Autoclavable | |
Plastic |