Cytiva Whatman™ 30mm Puradisc PTFE Syringe Filters
Designed to be resistant to organic solvents. Whatman™ 30mm Puradisc PTFE Syringe Filters are ideal for the clarification of aggressive organic solvents and can also be used as a venting filter for small vessels.
$498.00 - $2391.00
Specifications
Material PTFE
Housing Material Polypropylene (PP)
Inlet Style Female Luer lock
Outlet Style Male Luer
Type Syringe Filter
View More Specs
Products 8
Catalog Number Mfr. No. Color Code Pore Size Price Quantity  
Catalog Number Mfr. No. Color Code Pore Size Price Quantity  
09-302-180
View Documents Promotion Details
Cytiva
10463503
White 0.2 μm
Pack of 100 Each for $498.00
09-302-181
View Documents Promotion Details
Cytiva
10463505
White 0.2 μm
Pack of 500 Each for $2,391.00
09-302-182
View Documents Promotion Details
Cytiva
10463513
Green 0.45 μm
Pack of 100 Each for $498.00
09-302-183
View Documents Promotion Details
Cytiva
10463515
Green 0.45 μm
Pack of 500 Each for $2,362.00
09-302-184
View Documents Promotion Details
Cytiva
10463523
Yellow 1.0 μm
Pack of 100 Each for $498.00
09-302-185
View Documents Promotion Details
Cytiva
10463525
Yellow 1 μm
Pack of 500 Each for $2,384.00
09-302-186
View Documents Promotion Details
Cytiva
10463533
Gray 5 μm
Pack of 100 Each for $508.21
09-302-187
View Documents Promotion Details
Cytiva
10463535
Gray 5 μm
Pack of 500 Each for $2,384.00
Description
Description
Designed to be resistant to organic solvents. Whatman™ 30mm Puradisc PTFE Syringe Filters are ideal for the clarification of aggressive organic solvents and can also be used as a venting filter for small vessels.
  • Hydrophobic PTFE membrane is laminated with polypropylene
  • Optimal utilization of small sample volumes
  • Excellent chemical resistance against standard organic HPLC solvents
  • Polypropylene support
  • Used as a venting filter for small vessels
Specifications
Specifications
PTFE
Female Luer lock
Syringe Filter
Polypropylene (PP)
Male Luer
30 mm
SDS
missing translation for 'documents'
missing translation for 'documents'