missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Whatman™ Polyester Drain Discs

Provides a flat surface to eliminate filter tearing or rupturing. Whatman™ Polyester Drain Discs are used to support membrane filters. They are available in a variety of diameters for use in a range of devices.

$103.49 - $140.96

Specifications

Material Polyester
Thickness (Metric) 100 μm
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (Metric) Membrane Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Membrane Price Quantity  
05-715-116
View Documents Promotion Details
Cytiva
WHT230300
10 mm
Each for $103.49
 
09-927-164
View Documents Promotion Details
Cytiva
230500
22 mm
Pack of 100 Each for $110.00
 
09927167
View Documents Promotion Details
Cytiva
231100
47 mm
Pack of 100 Each for $140.96
 
09927165
View Documents Promotion Details
Cytiva
230800
37 mm
Pack of 100 Each for $121.16
 
Description

Description

Provides a flat surface to eliminate filter tearing or rupturing. Whatman™ Polyester Drain Discs are used to support membrane filters. They are available in a variety of diameters for use in a range of devices.

For use with membrane hardware where extra support is needed for improved flow rate and throughput. Can be used as a separator between membranes in serial stack filtration applications.

  • Chemically inert polyester
  • 100μm Thickness
  • Binder-free
  • Available in several diameters

Specifications

Specifications

Polyester
100 μm
SDS
missing translation for 'documents'

missing translation for 'documents'