QIAGEN OmniSwab, Sterile
For buccal and saliva samples.
Supplier: QIAGEN WB100035
Description
For buccal and saliva samples.
- Sterile collection swabs with ejectable brush-like head used for buccal and saliva samples and convenient DNA extraction.
- OmniSwabs are manufactured with an absorbent material that increases buccal cell concentrations to give high quality results.
- Compatible with standard STR amplification chemistry.
- OmniSwab collection swabs are individually packaged in a clean-room environment and sterilized by gamma irradiation.
Specifications
Ejectable head | |
For the collection of saliva and cheek buccal cells | |
Sterile |
Swab | |
Swab |