GE Healthcare Whatman™ Minifold I Dot Blot System
Well manifolds for proteins and nucleic acids
Manufacturer:  GE Healthcare 10447900
Catalog No. 36-100-5849
Includes: 96-well manifold apparatus, 5 × Protran BA85 sheets, 5 × 3mm Chr sheets
Description
Description
  • Consists of a sample well plate, filter support plate, vacuum plenum, and metal clamping plate
  • Tension on clamping is adjustable, permitting use of a variety of blotting and filtration media
  • Compatible with multichannel pipettes
  • Well plate is made from acrylic
  • Novel O-ring design ensures discrete dot formation without leakage of samples by cross lateral flow
  • Generates even, uniform dots that eliminate uneven test areas resulting from manual sample application
  • 12.5mm2 application area
  • 500μL/well
Specifications
Specifications
Minifold™ I dot-blot system
5.31 in.
7cm
11.5cm
5.31 x 4.5 x 2.75 in. (13.5 x 11.5 x 7cm)
Acrylic
2.75 in.
4.5 in.
13.5cm
Multichannel pipette
96-well Acrylic manifold apparatus (components-sample plate, filter support plate, vacuum plenum, clamping plate), protran BA85 sheets, 3mm chromatography sheets, well plate
Documents