missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/B
Filters fine particles with high wet-strength and high loading capacity. Whatman™ Binder-Free Microfiber Filters, Grade GF/B are particularly useful for liquid clarification and liquid scintillation counting.
$78.00 - $290.42
Specifications
Basis Weight | 143 g/m2 |
---|---|
Description | GEâs Whatman brand Grade GF/B is a thick filter with fine particle filter and good loading capacity. GF/B filters are particularly useful for liquid clarification and liquid scintillation counting. |
Format | Disc |
Material | Borosilicate Glass |
Particle Retention | 1.0 μm |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | |||||
09-874-20
|
Cytiva
1821021 |
21 mm |
Pack of 100 Each for $95.61
|
|
|||||
09-874-22
|
Cytiva
1821024 |
24 mm |
Pack of 100 Each for $108.33
|
|
|||||
09-874-22A
|
Cytiva
1821025 |
25 mm |
Pack of 100 Each for $112.88
|
|
|||||
09-874-22B
|
Cytiva
1821037 |
37 mm |
Pack of 100 Each for $131.20
|
|
|||||
09-874-24
|
Cytiva
1821042 |
42.5 mm |
Pack of 100 Each for $144.32
|
|
|||||
09-874-24A
|
Cytiva
1821047 |
47 mm |
Pack of 100 Each for $229.27
|
|
|||||
09-874-26
|
Cytiva
1821055 |
55 mm |
Pack of 100 Each for $183.68
|
|
|||||
09-874-26A
|
Cytiva
1821070 |
70 mm |
Pack of 100 Each for $244.00
|
|
|||||
09-874-26B
|
Cytiva
1821090 |
90 mm |
Pack of 25 Each for $78.00
|
|
|||||
09-874-26C
|
Cytiva
1821110 |
110 mm |
Pack of 25 Each for $121.84
|
|
|||||
09-874-26D
|
Cytiva
1821125 |
125 mm |
Pack of 25 Each for $123.67
|
|
|||||
09-874-26E
|
Cytiva
1821150 |
150 mm |
Pack of 25 Each for $290.42
|
|
|||||
09-927-765
|
Cytiva
1821185 |
185 mm |
Pack of 25 Each for $235.00
|
|
|||||
Description
Filters fine particles with high wet-strength and high loading capacity. Whatman™ Binder-Free Microfiber Filters, Grade GF/B Sheets are particularly useful for liquid clarification and liquid scintillation counting.
Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500ºC. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
- Fine particle filter with high wet-strength and high loading capacity
- Useful for liquid clarification and liquid scintillation counting
- Particularly useful when liquid clarification or solids quantification is required for heavily loaded fine particulate suspensions
- Used in LSC techniques where high loading capacity is required
- Can be used as a finely retentive membrane filter
Specifications
143 g/m2 | |
Disc | |
1.0 μm | |
Grade GF/B | |
500°C | |
675 μm | |
81 mL/min |
GEâs Whatman brand Grade GF/B is a thick filter with fine particle filter and good loading capacity. GF/B filters are particularly useful for liquid clarification and liquid scintillation counting. | |
Borosilicate Glass | |
5.5 sec./100mL | |
932°F | |
0.02 in. | |
Filter |