Learn More
Invitrogen™ Novex™ Tris-Glycine Mini Protein Gels, 16%, 1.0 mm, WedgeWell™ format
Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity.
Supplier: Invitrogen™ XP00165BOX
Description
Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity. Novex Tris-Glycine Mini Gels are based on traditional Laemmli protein electrophoresis that allow the use of Laemmli sample and running buffers.
Features of Novex Tris-Glycine gels, WedgeWell format, include:
• Improved shelf life—store gels for up to 16 weeks at 4°C
• Diversity—use for native and denaturing protein assays
• Wedge-shaped wells—easily load up to two times more sample volume
• Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes
Choose the right gel format for your experiments
Novex Tris-Glycine gels, WedgeWell format, come in a variety of fixed concentrations from 6% to 18%, as well as gradients with ranges of 4–12%, 4–20%, 8–16%, and 10–20%. You can select from our many well formats, including 10-, 12-, and 15-well, and different pack sizes.
Run your proteins in native or denatured form
Novex Tris-Glycine gels, WedgeWell format, do not contain SDS and can be used to run your proteins in native or in denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26755). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672).
Convenient to use
Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.
For transferring your proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) when preforming traditional wet transfer using the Invitrogen Mini Blot Module (B1000). Transfers can also be performed with rapid semi-dry transfer using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).
Specifications
Mini Gel Tank, XCell SureLock Mini-Cell | |
Mini | |
Tris-Glycine Plus | |
Up to 35 μL | |
Denaturing, Native | |
Molecular Weight | |
10 Gels/Box | |
Store at 2°C to 8°C. Do not freeze. | |
8 cm |
16% | |
1.0 mm | |
Denaturing, Native | |
6 to 200 kDa | |
15-well | |
Novex™, WedgeWell™ | |
Wet Ice | |
8 cm |