Invitrogen™ WedgeWell™ Tris-Glycine Mini Gels DFS Item
Polyacrylamide gels based on traditional Laemmli protein electrophoresis, which allows the use of Laemmli sample and running buffers
$37.22 - $161.00
Specifications
Length (Metric) Gel 8cm
Width (Metric) Gel 8cm
Material Tris-Glycine Gel
Thickness (Metric) 1.0mm
View More Specs
Products 33
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
XP04200BOX View Documents Invitrogen™
LSXP04200BOX
4-20% Tris-Glycine Mini Gels, 10-well 10 Gels Pack for $161.00
XP00100BOX View Documents Invitrogen™
LSXP00100BOX
10% Tris-Glycine Mini Gels, 10-well 10 Gels Pack for $161.00
XP04202BOX View Documents Invitrogen™
LSXP04202BOX
4-20% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP00062BOX View Documents Invitrogen™
LSXP00062BOX
6% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP00060BOX View Documents Invitrogen™
LSXP00060BOX
6% Tris-Glycine Mini Gels, 10-well 10 Gels Pack for $161.00
XP00065BOX View Documents Invitrogen™
LSXP00065BOX
6% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP00080BOX View Documents Invitrogen™
LSXP00080BOX
10 Gels Pack for $161.00
XP00082BOX View Documents Invitrogen™
LSXP00082BOX
8% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP00085BOX View Documents Invitrogen™
LSXP00085BOX
8% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP00100PK2 View Documents Invitrogen™
LSXP00100PK2
10% Tris-Glycine Mini Gels, 10-well, 2-pack 2 Gels Pack of 2 Each for $37.22
XP00102BOX View Documents Invitrogen™
LSXP00102BOX
10% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP00120BOX View Documents Invitrogen™
LSXP00120BOX
12% Tris-Glycine Mini Gels, 10-well, Box of 10 10 Gels Pack for $161.00
XP00122BOX View Documents Invitrogen™
LSXP00122BOX
12% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP00140BOX View Documents Invitrogen™
LSXP00140BOX
14% Tris-Glycine Mini Gels, 10-well 10 Gels Pack for $161.00
XP00125BOX View Documents Invitrogen™
LSXP00125BOX
12% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP00145BOX View Documents Invitrogen™
LSXP00145BOX
14% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP00142BOX View Documents Invitrogen™
LSXP00142BOX
14% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP00160BOX View Documents Invitrogen™
LSXP00160BOX
16% Tris-Glycine Mini Gels, 10-well 10 Gels Pack for $161.00
XP00162BOX View Documents Invitrogen™
LSXP00162BOX
16% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP00165BOX View Documents Invitrogen™
LSXP00165BOX
16% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP04120PK2 View Documents Invitrogen™
LSXP04120PK2
4-12% Tris-Glycine Mini Gels, 10-well, 2-pack 2 Gels Pack of 2 Each for $37.22
XP04120BOX View Documents Invitrogen™
LSXP04120BOX
4-12% Tris-Glycine Mini Gels, 10-well 10 Gels Pack for $161.00
XP04125BOX View Documents Invitrogen™
LSXP04125BOX
4-12% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP04122BOX View Documents Invitrogen™
LSXP04122BOX
4-12% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP04200PK2 View Documents Invitrogen™
LSXP04200PK2
2 Gels Pack of 2 Each for $37.22
XP04205BOX View Documents Invitrogen™
LSXP04205BOX
4-20% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP08160BOX View Documents Invitrogen™
LSXP08160BOX
8-16% Tris-Glycine Mini Gels, 10-well 10 Gels Pack for $161.00
XP08162BOX View Documents Invitrogen™
LSXP08162BOX
8-16% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP08165BOX View Documents Invitrogen™
LSXP08165BOX
8-16% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP10200BOX View Documents Invitrogen™
LSXP10200BOX
10-20% Tris-Glycine Mini Gels, 10-well 10 Gels Pack for $161.00
XP10202BOX View Documents Invitrogen™
LSXP10202BOX
10-20% Tris-Glycine Mini Gels, 12-well 10 Gels Pack for $161.00
XP10205BOX View Documents Invitrogen™
LSXP10205BOX
10-20% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
XP00105BOX View Documents Invitrogen™
LSXP00105BOX
10% Tris-Glycine Mini Gels, 15-well 10 Gels Pack for $161.00
Description
Description
Novex™ WedgeWell™ Tris-Glycine mini gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis, which allows the use of Laemmli sample and running buffers.
Novex WedgeWell Tris-Glycine gels offer reproducible separation of a wide range of proteins into well-resolved bands.
Features of the Novex WedgeWell gels include:
Improved shelf life—store gels for up to 12 months at 4°C
Diversity—use for native and denaturing protein assays
Wedge-shaped wells—easily load up to two times more sample volume
Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes
Choose the right gel format for your experiments
Novex WedgeWell Tris-Glycine gels come in a variety of fixed concentrations from 6% to 18%, as well as gradients with ranges of 4–12%, 4–20%, 8–16% and 10–20%. You can select from our many well formats, including 10-, 12- and 15-well, and different pack sizes.
For best results, most Novex Tris-Glycine gels should be stored at 4°C and used within 8 weeks of purchase. Novex 16% and 18% Tris-Glycine gels should be used within 4 weeks of purchase.
Run your proteins in native or denatured form
Novex WedgeWell Tris-Glycine gels do not contain SDS and can be used to run your proteins in native or in denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer and Novex Tris-Glycine SDS Running Buffer. For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer and Novex Tris-Glycine Native Running Buffer.
Convenient to use
Novex WedgeWell Tris-Glycine gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.
For transferring your proteins to membrane, we recommend using Novex Tris-Glycine Transfer Buffer. Transfers can be done using the Mini Blot Module or the iBlot Gel Transfer Device.
Specifications
Specifications
8cm
8cm
Tris-Glycine Gel
1.0mm
For Research Use Only. Not for use in diagnostic procedures.