missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, TraceSELECT™, For Trace Analysis, Honeywell Riedel-de Haën™
TraceSELECT™, for trace analysis
$121.93 - $1407.05
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
60047769
|
Honeywell-Riedel-de Haen
95305500ML |
500 mL | Poly Bottle |
Each for $121.93
|
|
||||
60047765
|
Honeywell Chemicals
953051L |
1 L | Glass Bottle |
Each for $173.65
|
|
||||
60047766
|
Honeywell Chemicals
9530525L |
2.5 L | Poly Bottle |
Each for $405.18
|
|
||||
60047764
|
Honeywell Chemicals
9530510L |
10 L | Cubitainer |
Each for $569.73
|
|
||||
60047768
|
Honeywell Chemicals
953054X25L |
4 x 2.5 L | Poly Bottle |
Case of 4 Each for $1,407.05
|
|
||||
Description
High purity solvents for speciation analysis by LC-ICP-MS
Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance
The blank values for metal traces in these solvents are in the ppb range or lower.
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
6 to 8 (at 20°C) | |
Colorless | |
100°C | |
MFCD00011332 | |
2050024 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02g/mol | |
Odorless | |
1g/mL at 20°C | |
Liquid |
7732-18-5 | |
0°C | |
H2O | |
NONH for all modes of transport | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
TraceSELECT™ | |
Poly Bottle | |
500 mL | |
Water |
Safety and Handling
RTECSNumber : 231-791-2