missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, LC/MS grade, Acros Organics™

Manufacturer:  Acros Organics AC615150025

Catalog No. AC615150025



Description

Description

ICP-OES/-MS standards
Specifications

Specifications

0.005 AU max. at 254nm, 0.005 AU max. at 260nm, 0.005 AU max. at 280nm, 0.01 AU max. at 210nm, 0.01 AU max. at 220nm, 0.01 AU max. at 230nm, 0.01 AU max. at 240nm
7732-18-5
10ppb max.
10ppb max.
10ppb max.
H2O
10ppb max.
dihydrogen oxide, dihydrogen monoxide
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
10ppb max.
50ppb max.
Pass Test (LC and LCMS)
10ppb max.
10ppb max.
10ppb max.
20ppb max.
10ppb max.
10ppb max.
1.0000g/mL
2.5L
Water
10ppb max.
20ppb max.
10ppb max.
H2O
MFCD00011332
15, 10237
Not detected
O
18.015
CHEBI:15377
18.02
50ppb max.
10ppb max.
10ppb max.
Protease free
1ppm max.
10ppb max.
17.5mmHg at 20°C
0.89mPa.s at 25°C
100°C

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
SDS Documents