missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, Deionized, ASTM Type I, Certified ACS Grade, LabChem™

$28.97 - $446.03

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC267402 View Documents LabChem
LC267402
1L Poly Bottle Each for $28.97
LC26740200 View Documents LabChem
LC26740200
200L Poly Drum Each for $446.03
LC267404 View Documents LabChem
LC267404
4L Poly Bottle Each for $36.26
LC267405 View Documents LabChem
LC267405
20L Cubitainer™ Each for $84.71
LC267406 View Documents LabChem
LC267406
10L Cubitainer™ Each for $62.31
Specifications

Chemical Identifiers

7732-18-5
18.015
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane

Specifications

7732-18-5
≤ 0.056uS/cm
H2O
Soluble in water
O
Passes Test
962
18.02
Certified ACS
0°C
0.99823g/mL
100°C
Water, Deionized, ASTM Type I
100
H2O
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
18.015
CHEBI:15377
Liquid
Poly Bottle
1.004cSt
Colorless
1L
Safety and Handling

Safety and Handling

GHS H Statement
Product is not hazardous.

EINECSNumber : 231-791-2

Recommended Storage : Room Temperature