missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, CHROMASOLV™, Honeywell Riedel-de Haën™
CHROMASOLV™, for pesticide residue analysis
$214.83 - $1769.38
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
7732-18-5 | |
MFCD00011332 | |
2050024 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02g/mol | |
1.000g/mL at 3.98°C | |
Water |
H2O | |
NONH for all modes of transport | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
CHROMASOLV™ | |
1 L |
Safety and Handling
RTECSNumber : ZC0110000