missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MKH4531 View Documents Avantor Performance
H45305
1L Poly N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MKH45320 View Documents Avantor Performance
H45309
20L Cubitainer N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane

Specifications

7732-18-5
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
ACS
1L
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Poly
Water