American Educational Products Watch Glass Form Covers
$61.06 - $61.06
Specifications
For Use With (Application) Used for covering boiling solutions
For Use With (Equipment) For 30 to 4000mL low form beaker, also for funnels, dishes, and jars
Shape Concave
View More Specs
Products 1 Description
Description
Concave shape prevents sliding. Returns condensation to beaker. Great for covering boiling solutions. For 30 to 4000mL low-form beakers. Also for funnels, dishes, and jars. Fire-polished edges for strength.
Specifications
Specifications
Used for covering boiling solutions
For 30 to 4000mL low form beaker, also for funnels, dishes, and jars
Concave