missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

W.S. TYLER™ RO-TAP™ Sieve Shakers

Recommended for accurate and consistent particle analysis testing

$3,794.33 - $9,050.10

Specifications

No. of Sieves 19.0
View More Specs

Requires: Sieves and receiver (order separately)


Products 3
Catalog Number Mfr. No. Diameter (English) Hertz Voltage Price Quantity  
Catalog Number Mfr. No. Diameter (English) Hertz Voltage Price Quantity  
04-909 Accessories View Documents W.S. TYLER™
RX29
8 in. 60Hz 110V Each for $3,794.33
04-909-3 View Documents W.S. TYLER™
RX30
12 in. 60Hz 110V Each for $4,306.50
04-909-2 View Documents W.S. TYLER™
RX2910*
8 in. 50Hz 220V Each for $9,050.10
Description

Description

Recommended for accurate and consistent particle analysis testing

The shaker has a unique two dimentional operation: a horizontal, circular motion and a vertical, topping motion which allow material particles to stratify and seek critical openings in the test media.

  • Heavy duty, stationary unit With digital 99 min. timer/clock, accurate to 0.1 second
  • Requires minimal maintenance
  • Each unit test run and serialized before shipment

Order Info

Available in 8 or 12 in. (20 or 30cm) diameters
Unit dimensions: 21L x 28W x 25 in. H (53 x 71 x 64cm)
Unit Weight: 180 lb. (82 kg)

Specifications

Specifications

19.0