W.S. TYLER™ RO-TAP™ Sieve Shakers
Recommended for accurate and consistent particle analysis testing
Supplier:  W.S. TYLER™ RX2910
 View more versions of this product
Catalog No. 04-909-2
Description
Description
Recommended for accurate and consistent particle analysis testing
The shaker has a unique two dimentional operation: a horizontal, circular motion and a vertical, topping motion which allow material particles to stratify and seek critical openings in the test media.
  • Heavy duty, stationary unit With digital 99 min. timer/clock, accurate to 0.1 second
  • Requires minimal maintenance
  • Each unit test run and serialized before shipment
Specifications
Specifications
220 V, 50 Hz
Sieve Shaker
21 in.
8 in.
64 cm
71 cm
86 kg
99 min.
220 V
50 Hz
54 cm
25 in.
28 in.
190 lb.
19