Learn More
Vinylene carbonate, 98%, stabilized, Thermo Scientific Chemicals
$241.12 - $241.12
Chemical Identifiers
CAS | 872-36-6 |
---|---|
Molecular Formula | C3H2O3 |
Molecular Weight (g/mol) | 86.05 |
MDL Number | MFCD00005380 |
InChI Key | VAYTZRYEBVHVLE-UHFFFAOYSA-N |
Synonym | vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3 |
PubChem CID | 13385 |
SMILES | O=C1OC=CO1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC140890050
|
Thermo Scientific Chemicals
140890050 |
5 g | Glass bottle |
Each for $241.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
872-36-6 | |
86.05 | |
VAYTZRYEBVHVLE-UHFFFAOYSA-N | |
13385 |
C3H2O3 | |
MFCD00005380 | |
vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3 | |
O=C1OC=CO1 |
Specifications
872-36-6 | |
Colorless to Yellow | |
162°C | |
Authentic | |
Glass bottle | |
1.4200 to 1.423 | |
5 g | |
13,342 | |
vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3 | |
VAYTZRYEBVHVLE-UHFFFAOYSA-N | |
2H-1,3-dioxol-2-one | |
0.1% max. BHT | |
86.05 | |
Liquid |
19°C to 22°C | |
1.3600g/mL | |
73°C | |
97.5% min. (GC) | |
C3H2O3 | |
MFCD00005380 | |
19, V,4, 72 | |
1.36 | |
Solubility in water: 11.5g / 100mL | |
O=C1OC=CO1 | |
86.05 | |
13385 | |
98% | |
Vinylene carbonate, Stabilized |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposu
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
EINECSNumber : 212-825-5
RUO – Research Use Only