Vileda Professional™ Telescopic Handle, short
For use in controlled environments for effective cleaning. Vileda Professional Telescopic Handle, short with adjustable length of 50 – 90cm.
Supplier: Vileda Professional™ 111384
Description
- Constructed of aluminum
- Adjustable length of 50 - 90cm
- Extends with twist of the wrist
- Anti-slip top grip to reduce slipping on walls
- Ergonomic grip allows blood circulation in hands and fingers, thereby reducing fatigue
- Resists autoclave, gamma and ETO processes
- Fits CE Squeegee and other commonly used frames
Specifications
Telescopic Handle, Short | |
Blue, Silver | |
Swep Duo Plus Frame | |
Floors, Walls and Ceilings, ISO 5, LAR, Production, Labs |