missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

VeriSpec™ Triethanolamine - HN(C2H4OH)3+ Standard for Ion Chromatography, 1000ppm in H2O, ISO 17025/Guide 34, Ricca Chemical

Formulated for high performance and reliable results

$180.97 - $351.88

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010948100 View Documents Ricca Chemical Company
RIRV010948100C
100mL Each for $180.97
RV010948250 View Documents Ricca Chemical Company
RIRV010948250C
250mL Each for $246.30
RV010948500 View Documents Ricca Chemical Company
RIRV010948500C
500mL Each for $351.88
Description

Description

  • 1000ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

102-71-6
149.19
7618
2-[bis(2-hydroxyethyl)amino]ethanol
C6H15NO3
GSEJCLTVZPLZKY-UHFFFAOYSA-N
CHEBI:28621
C(CO)N(CCO)CCO

Specifications

102-71-6,7732-18-5
100%,0.102%
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethanol
7618
Liquid
Bottle, Amber Glass
1.01
97.902%,0.098%
C6H15NO3
C(CO)N(CCO)CCO
149.19
CHEBI:28621
High Purity
100mL